COMH4211BBC5 Combi-Blocks, Inc.

1-(3-Aminopropyl)azetidin-3-ol

  • MDL number MFCD09745963
  • Cas Number : 1219979-33-5
  • Molecular Weight : 130.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6702

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6702
Availability details for
Quantity
  • Availability
  • Add to Cart