COMH4211BBC6 Combi-Blocks, Inc.

Methyl 4-(4-methoxybenzyl)itaconate

  • MDL number MFCD31618964
  • Cas Number : 130165-86-5
  • Molecular Weight : 264.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(4-methoxybenzyl) 1-methyl 2-methylenesuccinate
Inchi Code 1S/C14H16O5/c1-10(14(16)18-3)8-13(15)19-9-11-4-6-12(17-2)7-5-11/h4-7H,1,8-9H2,2-3H3
Inchi Key CXWOMESUUNORHK-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6703

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6703
Availability details for
Quantity
  • Availability
  • Add to Cart