COMH4211BBC7 Combi-Blocks, Inc.

2-[4-(2-Fluoro-4-methyl-benzyloxy)-phenyl]-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane

  • MDL number MFCD31583130
  • Molecular Weight : 342.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-((2-fluoro-4-methylbenzyl)oxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Inchi Code 1S/C20H24BFO3/c1-14-6-7-15(18(22)12-14)13-23-17-10-8-16(9-11-17)21-24-19(2,3)20(4,5)25-21/h6-12H,13H2,1-5H3
Inchi Key GCDPABFQKBGFCZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6704
Availability details for
Quantity
  • Availability
  • Add to Cart