COMH4211BBCA Combi-Blocks, Inc.

tert-Butyl 2-bromo-3-phenylpropionate

  • MDL number MFCD20484531
  • Cas Number : 75898-42-9
  • Molecular Weight : 285.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6711

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6711
Availability details for
Quantity
  • Availability
  • Add to Cart