COMH4211BBCB Combi-Blocks, Inc.

2-Amino-n-benzyl-5-fluoro-benzamide

  • MDL number MFCD11101528
  • Cas Number : 301527-71-9
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6713
Availability details for
Quantity
  • Availability
  • Add to Cart