COMH4211BBCD Combi-Blocks, Inc.

[4-(Tetrahydropyran-4-yloxy)phenyl]methanol

  • MDL number MFCD09064965
  • Cas Number : 892501-95-0
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6722
Availability details for
Quantity
  • Availability
  • Add to Cart