COMH4211BBCF Combi-Blocks, Inc.

5-(2-Methylpropoxy)-2-nitrobenzoic acid

  • MDL number MFCD09805677
  • Cas Number : 154606-14-1
  • Molecular Weight : 239.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6730

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6730
Availability details for
Quantity
  • Availability
  • Add to Cart