COMH4211BBD2 Combi-Blocks, Inc.

3-[1-(Dimethylamino)ethyl]phenol hydrochloride

  • MDL number MFCD01707504
  • Cas Number : 5441-61-2
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6735
Availability details for
Quantity
  • Availability
  • Add to Cart