COMH4211BBD5 Combi-Blocks, Inc.

(1-Cyclopropylethyl)[(4-fluorophenyl)methyl]amine

  • MDL number MFCD11145262
  • Cas Number : 1019481-51-6
  • Molecular Weight : 193.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6743

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6743
Availability details for
Quantity
  • Availability
  • Add to Cart