COMH4211BBD6 Combi-Blocks, Inc.

N-(4-Aminobutyl)-6-(trifluoromethyl)pyridine-3-carboxamide

  • MDL number MFCD31539521
  • Cas Number : 1840178-20-2
  • Molecular Weight : 261.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6746
Availability details for
Quantity
  • Availability
  • Add to Cart