COMH4211BBD9 Combi-Blocks, Inc.

1-(2-(4-Chlorophenyl)-2,2-difluoroethyl)piperidin-4-amine

  • MDL number MFCD31618977
  • Cas Number : 913574-92-2
  • Molecular Weight : 274.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6754
Availability details for
Quantity
  • Availability
  • Add to Cart