COMH4211BBDA Combi-Blocks, Inc.

[3-(4-Fluoro-phenyl)-isoxazol-4-yl]-methanol

  • MDL number MFCD30072588
  • Cas Number : 1159252-41-1
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6755

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6755
Availability details for
Quantity
  • Availability
  • Add to Cart