COMH4211BBDB Combi-Blocks, Inc.

3-Chloro-n-ethyl-1h-pyrazol-4-amine

  • MDL number MFCD31613763
  • Cas Number : 1704066-37-4
  • Molecular Weight : 145.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6758

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6758
Availability details for
Quantity
  • Availability
  • Add to Cart