COMH4211BBDC Combi-Blocks, Inc.

5-Phenylpentan-2-one

  • MDL number MFCD01024555
  • Cas Number : 2235-83-8
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6759

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6759
Availability details for
Quantity
  • Availability
  • Add to Cart