COMH4211BBDF Combi-Blocks, Inc.

1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propan-2-ol

  • MDL number MFCD22494847
  • Cas Number : 1248556-14-0
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6763

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6763
Availability details for
Quantity
  • Availability
  • Add to Cart