COMH4211BBE1 Combi-Blocks, Inc.

3-[(4-Fluorophenyl)sulfanyl]propan-1-ol

  • MDL number MFCD12179155
  • Cas Number : 133982-60-2
  • Molecular Weight : 186.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6765

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6765
Availability details for
Quantity
  • Availability
  • Add to Cart