COMH4211BBE2 Combi-Blocks, Inc.

4-Bromo-2-(ethanesulfonyl)aniline

  • MDL number MFCD21296040
  • Cas Number : 1496140-07-8
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6766

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6766
Availability details for
Quantity
  • Availability
  • Add to Cart