COMH4211BBE3 Combi-Blocks, Inc.

3-Ethyl-2,4-dioxo- 1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride

  • MDL number MFCD31381616
  • Cas Number : 1469764-14-4
  • Molecular Weight : 288.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-ethyl-2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride
Inchi Code 1S/C10H9ClN2O4S/c1-2-13-9(14)7-5-6(18(11,16)17)3-4-8(7)12-10(13)15/h3-5H,2H2,1H3,(H,12,15)
Inchi Key PLBAYLCVSGYWBO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6768

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6768
Availability details for
Quantity
  • Availability
  • Add to Cart