COMH4211BBE6 Combi-Blocks, Inc.

(4-(Cyclohexyloxy)phenyl)methanol

  • MDL number MFCD11167633
  • Cas Number : 13485-84-2
  • Molecular Weight : 206.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6772

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6772
Availability details for
Quantity
  • Availability
  • Add to Cart