COMH4211BBEA Combi-Blocks, Inc.

3-Bromo-5-ethanesulfonylamino-n,n-dimethyl-benzamide

  • MDL number MFCD31613978
  • Cas Number : 2270911-99-2
  • Molecular Weight : 335.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6781
Availability details for
Quantity
  • Availability
  • Add to Cart