COMH4211BBEC Combi-Blocks, Inc.

4-Chloro-2-dimethylaminomethylene-3-oxo-butyric acid ethyl ester

  • MDL number MFCD31381649
  • Cas Number : 1246616-68-1
  • Molecular Weight : 219.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6783
Availability details for
Quantity
  • Availability
  • Add to Cart