COMH4211BBED Combi-Blocks, Inc.

5-Benzenesulfonyl-indan-1-one

  • MDL number MFCD31579384
  • Cas Number : 201745-71-3
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6789

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6789
Availability details for
Quantity
  • Availability
  • Add to Cart