COMH4211BBF0 Combi-Blocks, Inc.

4-Bromo-2-methyl-n-(2,2,2-trifluoro-ethyl)-benzamide

  • MDL number MFCD31557870
  • Cas Number : 864725-41-7
  • Molecular Weight : 296.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6792
Availability details for
Quantity
  • Availability
  • Add to Cart