COMH4211BBF1 Combi-Blocks, Inc.

3-Amino-n-ethyl-5-methoxybenzamide

  • MDL number MFCD23719957
  • Cas Number : 1346245-36-0
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-N-ethyl-5-methoxybenzamide
Inchi Code 1S/C10H14N2O2/c1-3-12-10(13)7-4-8(11)6-9(5-7)14-2/h4-6H,3,11H2,1-2H3,(H,12,13)
Inchi Key QPXCBVHAGGPGCX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6795

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6795
Availability details for
Quantity
  • Availability
  • Add to Cart