COMH4211BBF3 Combi-Blocks, Inc.

tert-Butyl 4-(2-aminopyridin-3-yl)-2-fluorobenzoate

  • MDL number MFCD31579440
  • Cas Number : 1715033-51-4
  • Molecular Weight : 288.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6797
Availability details for
Quantity
  • Availability
  • Add to Cart