COMH4211BBF4 Combi-Blocks, Inc.

5-Amino-n-methoxy-2-methylbenzamide

  • MDL number MFCD20701807
  • Cas Number : 658085-47-3
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6798
Availability details for
Quantity
  • Availability
  • Add to Cart