COMH4211BBF6 Combi-Blocks, Inc.

tert-Butyl 4-(1h-indazol-5-yl)piperidine-1-carboxylate

  • MDL number MFCD12408339
  • Cas Number : 1158767-08-8
  • Molecular Weight : 301.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6806
Availability details for
Quantity
  • Availability
  • Add to Cart