COMH4211BBF7 Combi-Blocks, Inc.

Trans 3-(4-tert-butoxycarbonylamino-cyclohexyl)-2-methyl-propionic acid

  • MDL number MFCD31579316
  • Molecular Weight : 285.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6807
Availability details for
Quantity
  • Availability
  • Add to Cart