COMH4211BBF9 Combi-Blocks, Inc.

5-Bromo-2-iodo-n-methoxy-n-methylbenzamide

  • MDL number MFCD18839808
  • Cas Number : 842135-31-3
  • Molecular Weight : 370.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6810

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6810
Availability details for
Quantity
  • Availability
  • Add to Cart