COMH4211BBFA Combi-Blocks, Inc.

3-Ethoxy-5-(methoxycarbonyl)benzoic acid

  • MDL number MFCD24039307
  • Cas Number : 706792-00-9
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6811
Availability details for
Quantity
  • Availability
  • Add to Cart