COMH4211BBFC Combi-Blocks, Inc.

2-(4-Iodo-phenoxy)-phenylamine

  • MDL number MFCD11103649
  • Cas Number : 860677-85-6
  • Molecular Weight : 311.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6815

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6815
Availability details for
Quantity
  • Availability
  • Add to Cart