COMH4211BBFF Combi-Blocks, Inc.

1-[4-Fluoro-2-(2-methoxy-ethoxy)-phenyl]-ethanone

  • MDL number MFCD17286509
  • Cas Number : 474708-51-5
  • Molecular Weight : 212.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6821
Availability details for
Quantity
  • Availability
  • Add to Cart