COMH4211BC00 Combi-Blocks, Inc.

(4-(3-Fluoropropoxy)-2-methylphenyl)methanol

  • MDL number MFCD31628774
  • Cas Number : 1616507-85-7
  • Molecular Weight : 198.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-(3-fluoropropoxy)-2-methylphenyl)methanol
Inchi Code 1S/C11H15FO2/c1-9-7-11(14-6-2-5-12)4-3-10(9)8-13/h3-4,7,13H,2,5-6,8H2,1H3
Inchi Key NIMRGMSZNGGCBX-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6827

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6827
Availability details for
Quantity
  • Availability
  • Add to Cart