COMH4211BC05 Combi-Blocks, Inc.

3-Fluoro-4-iodobenzamide

  • MDL number MFCD18396921
  • Cas Number : 1261744-58-4
  • Molecular Weight : 265.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6839
Availability details for
Quantity
  • Availability
  • Add to Cart