COMH4211BC07 Combi-Blocks, Inc.

Dipyridamole tripiperidine

  • MDL number MFCD28016442
  • Cas Number : 16982-40-4
  • Molecular Weight : 484.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6842

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6842
Availability details for
Quantity
  • Availability
  • Add to Cart