COMH4211BC0A Combi-Blocks, Inc.

1-(4-Fluorobenzyl)-2,5-dihydro-1h-pyrrole

  • MDL number MFCD20257789
  • Cas Number : 954416-86-5
  • Molecular Weight : 177.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6845

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6845
Availability details for
Quantity
  • Availability
  • Add to Cart