COMH4211BC0B Combi-Blocks, Inc.

L-Lysine, n6-[4-(1,1-dimethylethoxy)-1,4-dioxobutyl]-n2-[(9h-fluoren-9-ylmethoxy)carbonyl]-

  • MDL number MFCD31381712
  • Cas Number : 1354905-64-8
  • Molecular Weight : 524.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6846

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6846
Availability details for
Quantity
  • Availability
  • Add to Cart