COMH4211BC12 Combi-Blocks, Inc.

5-Bromo-2-nitro-n-[(1r)-1-phenylethyl]aniline

  • MDL number MFCD28405827
  • Cas Number : 1416336-79-2
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6866

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6866
Availability details for
Quantity
  • Availability
  • Add to Cart