COMH4211BC13 Combi-Blocks, Inc.

(5-Bromo-2-chloro-pyridin-3-yl)-(4-methyl-piperazin-1-yl)-methanone

  • MDL number MFCD15504362
  • Cas Number : 1457225-01-2
  • Molecular Weight : 318.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6869
Availability details for
Quantity
  • Availability
  • Add to Cart