COMH4211BC16 Combi-Blocks, Inc.

4-Ethoxy-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD16852853
  • Cas Number : 1340229-02-8
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6872

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6872
Availability details for
Quantity
  • Availability
  • Add to Cart