COMH4211BC17 Combi-Blocks, Inc.

4-(3-Iodo-1h-indazol-5-yl)-piperidine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD28964381
  • Cas Number : 1383705-90-5
  • Molecular Weight : 427.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6875

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6875
Availability details for
Quantity
  • Availability
  • Add to Cart