COMH4211BC18 Combi-Blocks, Inc.

1-(Methylsulfonyl)piperidine-4-carbonitrile

  • MDL number MFCD11099946
  • Cas Number : 438630-74-1
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6877

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6877
Availability details for
Quantity
  • Availability
  • Add to Cart