COMH4211BC19 Combi-Blocks, Inc.

2-[2-Fluoro-4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-1-piperidin-1-yl-ethanone

  • MDL number MFCD31613950
  • Molecular Weight : 347.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6879
Availability details for
Quantity
  • Availability
  • Add to Cart