COMH4211BC1B Combi-Blocks, Inc.

2-Chloro-4-(2,2,2-trifluoro-ethoxy)-benzylamine

  • MDL number MFCD17256367
  • Cas Number : 1339896-37-5
  • Molecular Weight : 239.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6885

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6885
Availability details for
Quantity
  • Availability
  • Add to Cart