COMH4211BC1C Combi-Blocks, Inc.

5-Bromo-3-cyclohexylmethoxy-pyridin-2-ylamine

  • MDL number MFCD30729414
  • Molecular Weight : 285.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6886
Availability details for
Quantity
  • Availability
  • Add to Cart