COMH4211BC1E Combi-Blocks, Inc.

N-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]cyclobutanecarboxamide

  • MDL number MFCD31618963
  • Cas Number : 2246888-24-2
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6890

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6890
Availability details for
Quantity
  • Availability
  • Add to Cart