COMH4211BC20 Combi-Blocks, Inc.

3-([(tert-Butyldimethylsilyl)oxy]methyl)piperidine

  • MDL number MFCD31583094
  • Cas Number : 876147-50-1
  • Molecular Weight : 229.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6898
Availability details for
Quantity
  • Availability
  • Add to Cart