COMH4211BC21 Combi-Blocks, Inc.

Carbamic acid, [(4-methoxyphenyl)methoxy]-, 1,1-dimethylethyl ester

  • MDL number MFCD24468168
  • Cas Number : 154083-67-7
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl ((4-methoxybenzyl)oxy)carbamate
Inchi Code 1S/C13H19NO4/c1-13(2,3)18-12(15)14-17-9-10-5-7-11(16-4)8-6-10/h5-8H,9H2,1-4H3,(H,14,15)
Inchi Key KHWKNPWCIFIYOC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6902

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6902
Availability details for
Quantity
  • Availability
  • Add to Cart