COMH4211BC23 Combi-Blocks, Inc.

4-Amino-n,2-dimethylbenzamide

  • MDL number MFCD16117685
  • Cas Number : 912838-54-1
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6905
Availability details for
Quantity
  • Availability
  • Add to Cart