COMH4211BC24 Combi-Blocks, Inc.

Hexanoic acid, 6-[(3-iodobenzoyl)amino]-

  • MDL number MFCD03544663
  • Cas Number : 247140-51-8
  • Molecular Weight : 361.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6906

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6906
Availability details for
Quantity
  • Availability
  • Add to Cart